Anti-NFATC4 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA323878EA 564 EUR
ORIGTA323878
Anti-NFATC4 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:2000-5000; Western Blot: 1:500-2000

Type: Primary
Antigen: NFATC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR