Anti-EBI3 Rat Monoclonal Antibody (Unconjugated) [clone: 10J811]
Lieferant: Rockland Immunochemicals
ROCK210-501-B66SEA 219 EUR
ROCK210-501-B66S
Anti-EBI3 Rat Monoclonal Antibody (Unconjugated) [clone: 10J811]
Antikörper
Anti-EBI-3 antibody has been tested for use in ELISA, Western Blot, and Flow Cytometry. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Flow Cytometry: 1.5-3.0µg/10x6 cells; Western Blot: 1µg/ml;
Type: Primary
Antigen: EBI3
Clonality: Monoclonal
Clone: 10J811
Conjugation: Unconjugated
Epitope: Full length
Host: Rat
Isotype: IgG
Reactivity: Mouse