Anti-AKT1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-401SEA 219 EUR
ROCK100-401-401S
Anti-AKT1 Rabbit Polyclonal Antibody
Antikörper
This monoclonal antibody is suitable for ELISA, immunochemistry, immunoprecipitation and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500 - 1:3000; Immunochemistry: 20 µg/ml

Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR