Anti-DAT extracellular loop 2 Rabbit Polyclonal Antibody

Lieferant: Cayman Chemical
CAYM10009373-1EA 558 EUR
CAYM10009373-1
Anti-DAT extracellular loop 2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DAT extracellular loop 2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR