Anti-Kir2.3 Mouse Monoclonal Antibody [clone: S25-35]

CAYM13716-100EA 515 EUR
CAYM13716-100
Anti-Kir2.3 Mouse Monoclonal Antibody [clone: S25-35]
Antikörper
Type: Primary
Antigen: Kir2.3
Clonality: Monoclonal
Clone: S25-35
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR