Anti-IgG Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: Abnova
ABNOPAB10667EA 419 EUR
ABNOPAB10667
Anti-IgG Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Rabbit anti-horse IgG recognizes horse IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein.
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: F(ab')2
Host: Rabbit
Isotype:
Reactivity: Horse