Anti-JAK2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB7655EA 399 EUR
ABNOPAB7655
Anti-JAK2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of JAK2.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: JAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR