Anti-IgG Goat Monoclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abnova
ABNOPAB10823EA 364 EUR
ABNOPAB10823
Anti-IgG Goat Monoclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Goat anti-rabbit IgG recognizes rabbit IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Alkaline Phosphatase.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR