Anti-IgG Donkey Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Abnova
ABNOPAB10815EA 170 EUR
ABNOPAB10815
Anti-IgG Donkey Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Donkey anti-rabbit IgG recognizes rabbit IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR