Anti-IgG Rabbit Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: Abnova
ABNOPAB10802EA 370 EUR
ABNOPAB10802
Anti-IgG Rabbit Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Rabbit anti-pig IgG recognizes pig IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Rhodamine.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: F(c)
Host: Rabbit
Isotype:
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR