Anti-IgG Rabbit Monoclonal Antibody (Biotin)
Lieferant: Abnova
ABNOPAB10801EA 419 EUR
ABNOPAB10801
Anti-IgG Rabbit Monoclonal Antibody (Biotin)
Antikörper
Rabbit anti-pig IgG recognizes pig IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin.
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope: F(ab')2
Host: Rabbit
Isotype:
Reactivity: Pig