Anti-IgG Sheep Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Lieferant: Abnova
ABNOPAB10780EA 262 EUR
ABNOPAB10780
Anti-IgG Sheep Monoclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Sheep anti-mouse IgG recognizes mouse IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Rhodamine.
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Sheep
Isotype:
Reactivity: Mouse