Anti-IgG Rabbit Monoclonal Antibody (Biotin)

Lieferant: Abnova
ABNOPAB10778EA 419 EUR
ABNOPAB10778
Anti-IgG Rabbit Monoclonal Antibody (Biotin)
Antikörper
Rabbit anti-mouse IgG recognizes mouse IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Biotin.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope: F(c)
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR