Anti-IDO1 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB6880EA 518 EUR
ABNOPAB6880
Anti-IDO1 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of IDO1.

Recommended Dilutions: ELISA: 1:8000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 4-6 ?g/ml

Type: Primary
Antigen: IDO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR