Anti-CNR2 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB6877EA 518 EUR
ABNOPAB6877
Anti-CNR2 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of CNR2.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 2-3 ?g/ml

Type: Primary
Antigen: CNR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR