Anti-IgG Rabbit Monoclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Abnova
ABNOPAB10771EA 419 EUR
ABNOPAB10771
Anti-IgG Rabbit Monoclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Rabbit anti-mouse IgG recognizes mouse IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Alkaline Phosphatase.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(ab')2
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR