Anti-ACKR1 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB6873EA 518 EUR
ABNOPAB6873
Anti-ACKR1 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of DARC.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.1-0.3 ?g/ml; Immunohistochemistry: 3 ?g/ml

Type: Primary
Antigen: ACKR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR