Anti-IgG Rabbit Monoclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Abnova
ABNOPAB10770EA 419 EUR
ABNOPAB10770
Anti-IgG Rabbit Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Rabbit anti-mouse IgG recognizes mouse IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase.
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(ab')2
Host: Rabbit
Isotype:
Reactivity: Mouse