Anti-Pseudomonas marginalis Rat Polyclonal Antibody

ABNOPAB14744EA 793 EUR
ABNOPAB14744
Anti-Pseudomonas marginalis Rat Polyclonal Antibody
Rat polyclonal antibody raised against Pseudomonas marginalis.

Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:1000-1:5000

Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: Pseudomonas marginalis
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR