Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 15B3]

Lieferant: Abnova
ABNOMAB5233EA 1250 EUR
ABNOMAB5233
Anti-TNFRSF8 Mouse Monoclonal Antibody [clone: 15B3]
Mouse monoclonal antibody raised against recombinant TNFRSF8.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:20-1:40

Type: Primary
Antigen: TNFRSF8
Clonality: Monoclonal
Clone: 15B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR