Anti-CD175 Mouse Monoclonal Antibody [clone: BRIC111]

Lieferant: Abnova
ABNOMAB5231EA 904 EUR
ABNOMAB5231
Anti-CD175 Mouse Monoclonal Antibody [clone: BRIC111]
Antikörper
Mouse monoclonal antibody raised against native CD175.

Type: Primary
Antigen: CD175
Clonality: Monoclonal
Clone: BRIC111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR