Anti-CD40L Mouse Monoclonal Antibody [clone: TRAP1.3.6]

Lieferant: Abnova
ABNOMAB5229EA 904 EUR
ABNOMAB5229
Anti-CD40L Mouse Monoclonal Antibody [clone: TRAP1.3.6]
Antikörper
Mouse monoclonal antibody raised against native CD40LG.

Recommended Dilutions: Flow Cytometry: 10 ?l of the s?ggested working dilution (1:10-1:50 to label 106 cells in 100 ?l

Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: TRAP1.3.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR