Anti-HSF1 Rabbit Polyclonal Antibody

ABNOPAB4339EA 351 EUR
ABNOPAB4339
Anti-HSF1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of HSF1.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR