Anti-ITGA1 Mouse Monoclonal Antibody [clone: TS2/7]
Lieferant: Abnova
ABNOMAB5224EA 1160 EUR
ABNOMAB5224
Anti-ITGA1 Mouse Monoclonal Antibody [clone: TS2/7]
Antikörper
Mouse monoclonal antibody raised against native ITGA1.
Recommended Dilutions: Flow Cytometry: 5 ?l of the antibody (neat) to label 10^6 cells
Type: Primary
Antigen: ITGA1
Clonality: Monoclonal
Clone: TS2/7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human