Anti-Nicotinamide (Vitamin PP) Polyclonal Antibody

ABNOPAB14734EA 907 EUR
ABNOPAB14734
Anti-Nicotinamide (Vitamin PP) Polyclonal Antibody
Rat polyclonal antibody raised against Nicotinamide (Vitamin PP).

Recommended Dilutions: ELISA: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Recommended Dilutions: ELISA: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: Nicotinamide (Vitamin PP)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR