Anti-CD55 Mouse Monoclonal Antibody [clone: 67]
Lieferant: Abnova
ABNOMAB5221EA 643 EUR
ABNOMAB5221
Anti-CD55 Mouse Monoclonal Antibody [clone: 67]
Antikörper
Mouse monoclonal antibody raised against native CD55.
Recommended Dilutions: Flow Cytometry: 1:10-1:25; Immunohistochemistry (Frozen sections): 1:100-1:1000
Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: 67
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human