Anti-Picolinic acid Polyclonal Antibody

ABNOPAB14728EA 907 EUR
ABNOPAB14728
Anti-Picolinic acid Polyclonal Antibody
Rabbit polyclonal antibody raised against Picolinic acid..

Recommended Dilutions: ELISA: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Recommended Dilutions: ELISA: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: Picolinic acid
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR