Anti-DOK5 Rabbit Polyclonal Antibody

ABNOPAB3409EA 351 EUR
ABNOPAB3409
Anti-DOK5 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of DOK5.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100

Type: Primary
Antigen: DOK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR