Anti-DOK4 Rabbit Polyclonal Antibody

ABNOPAB3408EA 351 EUR
ABNOPAB3408
Anti-DOK4 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of DOK4.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-200; Immunohistochemistry: 1:50-100

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-200; Immunohistochemistry: 1:50-100

Type: Primary
Antigen: DOK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR