Anti-PD1 Mouse Monoclonal Antibody [clone: 12A7D7]

Lieferant: Abnova
ABNOMAB8089EA 643 EUR
ABNOMAB8089
Anti-PD1 Mouse Monoclonal Antibody [clone: 12A7D7]
Antikörper
Mouse monoclonal antibody raised against partial recombinant Pdcd1.

Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: PD1
Clonality: Monoclonal
Clone: 12A7D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR