Anti-IL33 Monoclonal Antibody [clone: 12H6D12]

Lieferant: Abnova
ABNOMAB8087EA 440 EUR
ABNOMAB8087
Anti-IL33 Monoclonal Antibody [clone: 12H6D12]
Antikörper
Mouse monoclonal antibody raised against recombinant IL33.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: IL33
Clonality: Monoclonal
Clone: 12H6D12
Conjugation: Unconjugated
Epitope:
Host:
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR