Anti-CRP Mouse Monoclonal Antibody (Biotin) [clone: C7]
Lieferant: Abnova
ABNOMAB5101EA 419 EUR
ABNOMAB5101
Anti-CRP Mouse Monoclonal Antibody (Biotin) [clone: C7]
Antikörper
Mouse monoclonal antibody raised against CRP.
Type: Primary
Antigen: CRP
Clonality: Monoclonal
Clone: C7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human