Anti-NT5E Mouse Monoclonal Antibody [clone: 1D7]

Lieferant: Abnova
ABNOMAB10838EA 440 EUR
ABNOMAB10838
Anti-NT5E Mouse Monoclonal Antibody [clone: 1D7]
Antikörper
Mouse monoclonal antibody raised against partial recombinant NT5E.

Recommended Dilutions: ELISA: 1:10000; Immunohistochemistry: 1:200-1:1000

Type: Primary
Antigen: NT5E
Clonality: Monoclonal
Clone: 1D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR