Anti-RIF1 Goat Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25455EA 440 EUR
ABNOPAB25455
Anti-RIF1 Goat Polyclonal Antibody
Antikörper
Goat polyclonal antibody raised against synthetic peptide of RIF1.

Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: RIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR