Anti-ADGRB3 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25530EA 545 EUR
ABNOPAB25530
Anti-ADGRB3 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of BAI3.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 4 ?g/ml

Type: Primary
Antigen: ADGRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Gorilla, Horse, Human, Monkey, Mouse, Rabbit, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR