Anti-IGSF8 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25604EA 545 EUR
ABNOPAB25604
Anti-IGSF8 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of IGSF8.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 15 ?g/ml

Type: Primary
Antigen: IGSF8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Gorilla, Human, Monkey, Pig, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR