Anti-ERAP1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25610EA 545 EUR
ABNOPAB25610
Anti-ERAP1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of ERAP1.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10-15 ?g/ml

Type: Primary
Antigen: ERAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Hamster, Human, Orangutan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR