Anti-SGSM3 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25623EA 440 EUR
ABNOPAB25623
Anti-SGSM3 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of SGSM3.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: SGSM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR