Anti-SAMSN1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25682EA 440 EUR
ABNOPAB25682
Anti-SAMSN1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of SAMSN1.

Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: SAMSN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR