Anti-BOK Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25702EA 440 EUR
ABNOPAB25702
Anti-BOK Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of BOK.

Recommended Dilutions: Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: BOK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR