Anti-MYBPC1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25710EA 440 EUR
ABNOPAB25710
Anti-MYBPC1 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of MYBPC1.

  • Recommended Dilutions: Immunohistochemistry: 5 μg/ml

Type: Primary
Antigen: MYBPC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR