Anti-SPINK7 Chicken Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25733EA 440 EUR
ABNOPAB25733
Anti-SPINK7 Chicken Polyclonal Antibody
Antikörper
Chicken polyclonal antibody raised against synthetic peptide of SPINK7.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: SPINK7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR