Anti-CASKIN2 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB25784EA 440 EUR
ABNOPAB25784
Anti-CASKIN2 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against synthetic peptide of CASKIN2.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 2.5 ?g/ml

Type: Primary
Antigen: CASKIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR