Anti-IgM Goat Monoclonal Antibody

Lieferant: Abnova
ABNOPAB9337EA 223 EUR
ABNOPAB9337
Anti-IgM Goat Monoclonal Antibody
Antikörper
Goat anti-mouse IgG recognizes mouse IgG IgA IgM whole molecules. This secondary antibody was purified using antigen affinity chromatography.

Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR