Anti-FKBP4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: Hi52C]

Lieferant: Abnova
ABNOMAB11382EA 545 EUR
ABNOMAB11382
Anti-FKBP4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: Hi52C]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of FKBP4.

Recommended Dilutions: Western Blot: 1:2000; Immunohistochemistry: 1:250; Immunoprecipitation: 5 ?g with 10-20 ?l Protein A beads

Type: Primary
Antigen: FKBP4
Clonality: Monoclonal
Clone: Hi52C
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Dog, Hamster, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR