Anti-PECAM1 Mouse Monoclonal Antibody [clone: JC/70A]
Lieferant: Abnova
ABNOMAB11349EA 471 EUR
ABNOMAB11349
Anti-PECAM1 Mouse Monoclonal Antibody [clone: JC/70A]
Antikörper
Mouse monoclonal antibody raised against recombinant PECAM1.
Recommended Dilutions: Flow Cytometry: 0.5-1 ?g/million cells; Immunohistochemistry: 1:100-1:200 for 30 min at RT; Western Blot: 1:200-1:400
Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: JC/70A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human