Anti-TIMM9 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOPAB28616EA 545 EUR
ABNOPAB28616
Anti-TIMM9 Rabbit Polyclonal Antibody
Antikörper
Rabbit polyclonal antibody raised against recombinant TIMM9.

Recommended Dilutions: Immunohistochemistry: 1:10-1:20; Western Blot: 1:100-1:250; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: TIMM9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR