Anti-CK17 Mouse Monoclonal Antibody [clone: E3]
Lieferant: Abnova
ABNOMAB11306EA 471 EUR
ABNOMAB11306
Anti-CK17 Mouse Monoclonal Antibody [clone: E3]
Antikörper
Mouse monoclonal antibody raised against KRT17.
Recommended Dilutions: Western Blot: 1:200 -1:2000; Flow Cytometry: 1:10 - 1:1000; Immunocytochemistry/Immunofluorescence: 1:10 - 1:2000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:50-1:100
Type: Primary
Antigen: CK17
Clonality: Monoclonal
Clone: E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human