Anti-BCL2 Mouse Monoclonal Antibody [clone: 100/D5]

Lieferant: Abnova
ABNOMAB11293EA 440 EUR
ABNOMAB11293
Anti-BCL2 Mouse Monoclonal Antibody [clone: 100/D5]
Antikörper
Mouse monoclonal antibody raised against synthetic peptide of BCL2.

Recommended Dilutions: Western Blot: 1-3 ?g/ml; Flow Cytometry: 0.5 ?g/l06 cells; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:250-1:1500; Immunoprecipitation: 1:10 - 1:500

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: 100/D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR