Anti-SLC24A1 Mouse Monoclonal Antibody [clone: 1E12]

Lieferant: Abnova
ABNOH00009187-M01EA 394 EUR
ABNOH00009187-M01
Anti-SLC24A1 Mouse Monoclonal Antibody [clone: 1E12]
Antikörper
Mouse monoclonal antibody raised against a partial recombinant SLC24A1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SLC24A1
Clonality: Monoclonal
Clone: 1E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR